Learn More
Alfa Aesar™ 4'-Aminoacetanilide, 98+%
Product Code. 11415606
Change view
Click to view available options
Quantity:
25g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11415606 25g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11415606 Supplier Alfa Aesar™ Supplier No. B20740.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
CAS 122-80-5
Boiling Point 267°C
Flash Point 195°C (383°F)
Molecular Formula C8H10N2O
MDL Number MFCD00007853
Quantity 25g
Beilstein 742888
Sensitivity Air Sensitive
Merck Index 14,411
Synonym 4'-aminoacetanilide, n-4-aminophenyl acetamide, p-aminoacetanilide, 4-aminoacetanilide, 4-acetamidoaniline, p-amino acetanilide, p-acetamidoaniline, acetparamin, acetamide, n-4-aminophenyl, fourrine a
InChI Key CHMBIJAOCISYEW-UHFFFAOYSA-N
SMILES CC(=O)NC1=CC=C(C=C1)N
IUPAC Name N-(4-aminophenyl)acetamide
Molecular Weight (g/mol) 150.181
PubChem CID 31230
Formula Weight 150.18
Percent Purity ≥98%
Chemical Name or Material 4'-Aminoacetanilide