Learn More
4-Amino-6-hydroxypyrimidine, 98%, Thermo Scientific™
Product Code. 11386666
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11386666 5 g 5g
11396666 250 mg 250mg
11376666 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11386666 Supplier Thermo Scientific Alfa Aesar Supplier No. L03825.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1193-22-2
Molecular Formula C4H5N3O
Molecular Weight (g/mol) 111.10
MDL Number MFCD00051502
InChI Key HFMLLTVIMFEQRE-UHFFFAOYSA-N
Synonym 4-amino-6-hydroxypyrimidine, 6-aminopyrimidin-4-ol, 4-hydroxy-6-aminopyrimidine, 4-pyrimidinol, 6-amino, 4 1h-pyrimidinone, 6-amino, 6-aminopyrimidin-4 3h-one, 4-oxy-6-aminopyrimidine, 6-amino-4-pyrimidinol, 6-amino-3h-pyrimidin-4-one, 6-amino-4 1h-pyrimidinone
PubChem CID 70944
SMILES NC1=CC(=O)N=CN1
Specifications
Melting Point 262°C to 264°C
Quantity 5 g
Beilstein 112429
IUPAC Name 6-amino-1H-pyrimidin-4-one
Formula Weight 111.1
Percent Purity 98%
Chemical Name or Material 4-Amino-6-hydroxypyrimidine