Learn More
4-Amino-4'-nitrostilbene-2,2'-disulfonic Acid Disodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Amino-4'-nitrostilbene-2,2'-disulfonic Acid, Disodium Salt |
| CAS | 6634-82-8 |
| Molecular Formula | C14 H10 N2 O8 S2 . 2 Na |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C14H12N2O8S2.2Na/c15-11-5-3-9(13(7-11)25(19,20)21)1-2-10-4-6-12(16(17)18)8-14(10)26(22,23)24;;/h1-8H,15H2,(H,19,20,21)(H,22,23,24);;/q;2*+1/p-2/b2-1+;; |
| SMILES | [Na+].[Na+].Nc1ccc(\C=C\c2ccc(cc2S(=O)(=O)[O-])[N+](=O)[O-])c(c1)S(=O)(=O)[O-] |
| IUPAC Name | disodium;5-amino-2-[(E)-2-(4-nitro-2-sulfonatophenyl)ethenyl]benzenesulfonate |
| Molecular Weight (g/mol) | 444.35 |
| Formula Weight | 443.9674 |