Learn More
4-Acetylbenzonitrile, 98+%
Product Code. 10696584 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10696584 100 g Glass bottle 100g
10377490 5 g Glass bottle 5g
10163990 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10696584 Supplier Thermo Scientific Acros Supplier No. 167581000
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
CAS: 1443-80-7 | C9H7NO | 145.16 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1443-80-7
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00001825
InChI Key NLPHXWGWBKZSJC-UHFFFAOYSA-N
Synonym 4'-cyanoacetophenone, benzonitrile, 4-acetyl, p-cyanoacetophenone, 4-cyanoacetophenone, p-acetylbenzonitrile, 4-acetyl-benzonitrile, benzonitrile, p-acetyl, 4-acetylbenzenecarbonitrile, 4-acetylbenznitrile
PubChem CID 74044
IUPAC Name 4-acetylbenzonitrile
SMILES CC(=O)C1=CC=C(C=C1)C#N
Specifications
CAS Min % 98.0
CAS Max % 100.0
Melting Point 55°C to 58°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3COC6H4CN
Quantity 100 g
Beilstein 10, 695
Formula Weight 145.16
Percent Purity 98+%
Physical Form Crystalline Mass or Crystalline Powder and Chunks
Chemical Name or Material 4-Acetylbenzonitrile
Show More Show Less
compliance-icons
Product Identifier
  • 4-Acetylbenzonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.