Learn More
4-Acetyl Biphenyl, Mikromol
Product Code. 30461124
Click to view available options
Quantity:
25 mg
4 x 25 mg
Unit Size:
100mg
25mg
Product Code. 30461124
Brand: Mikromol MM3501.0100254AMP
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-Acetyl Biphenyl, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-acetyl biphenyl
CAS 92-91-1
Analyte or Component Names 4-acetyl biphenyl
Shipping Condition Room Temperature
Molecular Formula C14 H12 O
Certifications/Compliance ISO 17025
Synonym Felbinac Imp. A (EP), (1-[1,1'-Biphenyl]-4-yl)ethanone, 1,1'-Biphenyl-4-yl methyl ketone, 1-(4-Phenylphenyl)ethanone, 1-Acetyl-4-phenylbenzene, 1-Biphenyl-4-ylethanone, 4-Acetyl-1,1'-biphenyl, 4-Acetylbiphenyl, 4-Biphenylyl methyl ketone, 4-Ethanoylbiphenyl, 4'-Phenylacetophenone, Methyl 4-biphenylyl ketone, NSC 1875, p-Acetylbiphenyl, p-Phenylacetophenone, Felbinac Impurity A
InChI Formula InChI=1S/C14H12O/c1-11(15)12-7-9-14(10-8-12)13-5-3-2-4-6-13/h2-10H,1H3
Solution Type Neat
SMILES CC(=O)c1ccc(cc1)c2ccccc2
IUPAC Name 1-(4-phenylphenyl)ethanone
Molecular Weight (g/mol) 196.24
Quantity 4 x 25 mg
Formula Weight 196.0888 g/mol