Learn More
4-Acetaminophen-d3 Sulfate, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Acetaminophen-d3 Sulfate |
| CAS | 1020718-78-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C8 2H3 H6 N O5 S |
| Quantity | 10 mg |
| Synonym | Acetamide-2,2,2-d3, N-[4-(sulfooxy)phenyl]- (ACI), N-[4-(Sulfooxy)phenyl]acetamide-2,2,2-d3 (ACI), 4-Acetaminophen-d3 sulfate, Acetaminophen sulfate-d3, N-(Acetyl-d3)-4-aminophenol O-sulfate, Paracetamol O-sulfate D3 (acetyl-D3), Paracetamol-d3 sulfate |
| InChI Formula | InChI=1S/C8H9NO5S/c1-6(10)9-7-2-4-8(5-3-7)14-15(11,12)13/h2-5H,1H3,(H,9,10)(H,11,12,13)/i1D3 |
| SMILES | [2H]C([2H])([2H])C(=O)Nc1ccc(OS(=O)(=O)O)cc1 |
| IUPAC Name | [4-[(2,2,2-trideuterioacetyl)amino]phenyl] hydrogen sulfate |
| Molecular Weight (g/mol) | 234.24 |
| Show More |