missing translation for 'onlineSavingsMsg'
Learn More

4-(9-Carbazolyl)benzeneboronic acid pinacol ester, 95%, Thermo Scientific™

Catalog Number p-7034278
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
785051-54-9
C24H24BNO2
369.271
MFCD16294549
AHDSYMVAUJZCOP-UHFFFAOYSA-N
9-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-9h-carbazole, 9-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl-9h-carbazole, 9-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbazole, amtb250, 9-carbazolylbenzeneboronic acid pinacol ester, 4-9h-carbazol-9-yl phenylboronic acid pinacol ester, 9h-carbazole-9-4-phenyl boronic acid pinacol ester, 4-9h-carbazol-9-yl phenylboronic acid, pinacol ester
11850073
9-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbazole
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N3C4=CC=CC=C4C5=CC=CC=C53
This item is not returnable. View return policy
785051-54-9
C24H24BNO2
369.271
MFCD16294549
AHDSYMVAUJZCOP-UHFFFAOYSA-N
9-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-9h-carbazole, 9-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl-9h-carbazole, 9-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbazole, amtb250, 9-carbazolylbenzeneboronic acid pinacol ester, 4-9h-carbazol-9-yl phenylboronic acid pinacol ester, 9h-carbazole-9-4-phenyl boronic acid pinacol ester, 4-9h-carbazol-9-yl phenylboronic acid, pinacol ester
11850073
9-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbazole
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N3C4=CC=CC=C4C5=CC=CC=C53

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Air sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Quantity 1 g
Beilstein 10465891
Sensitivity Air Sensitive
Formula Weight 369.27
Percent Purity 95%
Chemical Name or Material 4-(9-Carbazolyl)benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'