Learn More
4-[6-(Tributylstannyl)-2-pyridinyl]morpholine, 95%, Thermo Scientific™
Product Code. 10715213
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10715213 1 g Amber Glass Bottle 1g
10012264 250 mg Amber Glass Bottle 250mg
11350607 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10715213 Supplier Thermo Scientific Maybridge Supplier No. CC38341DA
 Restricted Item
Estimated Shipment: 23-11-2026
Chemical Identifiers
CAS 869901-24-6
Molecular Formula C21H38N2OSn
Molecular Weight (g/mol) 453.258
MDL Number MFCD08271894
InChI Key LDERBIPTTICDSG-UHFFFAOYSA-N
Synonym 4-6-tributylstannyl pyridin-2-yl morpholine, 4-6-tributylstannyl-2-pyridinyl morpholine, pubchem21425, 4-6-tributylstannanyl-pyridin-2-yl-morpholine, morpholine,4-6-tributylstannyl-2-pyridinyl
PubChem CID 18525739
IUPAC Name tributyl-(6-morpholin-4-ylpyridin-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC=CC(=N1)N2CCOCC2
Specifications
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 453.25
Percent Purity 95%
Chemical Name or Material 4-[6-(Tributylstannyl)-2-pyridinyl]morpholine