Learn More
4-(6-Nitro-3-pyridinyl)-piperazine-2,2,6,6-d4, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(6-Nitro-3-pyridinyl)-piperazine-2,2,6,6-d4 |
| CAS | 2089333-08-2 |
| Molecular Formula | C9 H9 F3 N2 O2 |
| Quantity | 100 mg |
| Synonym | 1-(6-Nitro-3-pyridinyl)piperazine-d4, 1-(6-Nitropyridin-3-yl)piperazine-d4 |
| InChI Formula | InChI=1S/C9H9F3N2O2/c1-3-16-7(15)6-4-13-8(9(10,11)12)14-5(6)2/h4H,3H2,1-2H3 |
| SMILES | CCOC(=O)c1cnc(nc1C)C(F)(F)F |
| IUPAC Name | ethyl 4-methyl-2-(trifluoromethyl)pyrimidine-5-carboxylate |
| Molecular Weight (g/mol) | 234.175 |
| Formula Weight | 234.062 |