Learn More
4,6-Dimethylisophthalic acid, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 2790-09-2 |
| Molecular Formula | C10H10O4 |
| MDL Number | MFCD00230851 |
| Quantity | 100 mg |
| SMILES | O=C(C1=C(C)C=C(C)C(C(O)=O)=C1)O |
| Molecular Weight (g/mol) | 194.18 |
| Percent Purity | 98% |
| Chemical Name or Material | 4,6-Dimethylisophthalic acid |