Learn More
4,6-Dimethoxy-2-(methylsulfonyl)pyrimidine, TRC
Specifications
Specifications
| Chemical Name or Material | 4,6-Dimethoxy-2-(methylsulfonyl)pyrimidine |
| CAS | 113583-35-0 |
| Molecular Formula | C7H10N2O4S |
| Quantity | 10 g |
| Synonym | 4,6-Dimethoxy-2-(methylsulfonyl)-pyrimidine |
| InChI Formula | InChI=1S/C7H10N2O4S/c1-12-5-4-6(13-2)9-7(8-5)14(3,10)11/h4H,1-3H3 |
| SMILES | O=[S](C1=NC(OC)=CC(OC)=N1)(C)=O |
| IUPAC Name | 4,6-dimethoxy-2-(methylsulfonyl)pyrimidine |
| Molecular Weight (g/mol) | 218.23 |
| Formula Weight | 218.08 |