Learn More
4,6-Dimethoxy-2-(methylsulfonyl)pyrimidine, 96%
Product Code. 11451650
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11451650 1 g 1g
11461650 5 g 5g
11471650 25 g 25g
3 options
Product Code. 11451650
Brand: Thermo Scientific Alfa Aesar H27003.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 113583-35-0 | C7H10N2O4S | 218.23 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 113583-35-0
Molecular Formula C7H10N2O4S
Molecular Weight (g/mol) 218.23
MDL Number MFCD00672151
InChI Key ITDVJJVNAASTRS-UHFFFAOYSA-N
Synonym 4,6-dimethoxy-2-methylsulfonyl pyrimidine, 2-methanesulfonyl-4,6-dimethoxypyrimidine, 2-methylsulfonyl-4,6-dimethoxypyrimidine, 2-methylsulfony-4,6-dimethoxypyridine, 4,6-dimethoxy-2-methylsulfonyl-pyrimidine, 2-methanesulfonyl-4,6-dimethoxy-pyrimidine, 4,6-dimethoxy-2-methylsulphonyl pyrimidine, pyrimidine, 4,6-dimethoxy-2-methylsulfonyl, pubchem23351, maybridge1_008822
PubChem CID 838363
IUPAC Name 4,6-dimethoxy-2-methylsulfonylpyrimidine
SMILES COC1=CC(OC)=NC(=N1)S(C)(=O)=O
Specifications
Melting Point 129°C to 133°C
Quantity 1 g
Formula Weight 218.23
Percent Purity 96%
Chemical Name or Material 4,6-Dimethoxy-2-(methylsulfonyl)pyrimidine