Learn More
4,6-Dichloropyrimidine, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 4,6-Dichloropyrimidine |
| CAS | 1193-21-1 |
| Molecular Formula | C4 H2 Cl2 N2 |
| Quantity | 10 g |
| InChI Formula | InChI=1S/C4H2Cl2N2/c5-3-1-4(6)8-2-7-3/h1-2H |
| SMILES | Clc1cc(Cl)ncn1 |
| IUPAC Name | 4,6-dichloropyrimidine |
| Molecular Weight (g/mol) | 148.98 |
| Formula Weight | 147.9595 |