Learn More
(4,5-Dimethoxy-2-nitrophenyl)methanol, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 1016-58-6 |
| Molecular Formula | C9H11NO5 |
| MDL Number | MFCD00014701 |
| Quantity | 25 g |
| SMILES | COC1=CC(CO)=C(C=C1OC)[N+]([O-])=O |
| Molecular Weight (g/mol) | 213.19 |
| Percent Purity | 98% |
| Chemical Name or Material | (4,5-Dimethoxy-2-nitrophenyl)methanol |