missing translation for 'onlineSavingsMsg'
Learn More

4-(4-methylpiperazin-1-yl)phenylboronic acid, 97%, Maybridge

Catalog Number 10397534
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 747413-21-4
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C17H27BN2O2
MDL Number MFCD06795650
Quantity 1 g
Synonym 1-methyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazine, 4-4-methylpiperazin-1-yl phenylboronic acid pinacol ester, 4-4-methylpiperazin-1-yl phenylboronic acid, pinacol ester, 4-4-methyl-1-piperazinyl phenylboronic acid pinacol ester, 4-4-methylpiperazin-1-yl benzeneboronic acid, pinacol ester, 1-methyl-4-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazine, 4-4-methyl-1-piperazinyl benzeneboronic acid pinacol ester, 2-4-4-methylpiperazin-1-yl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, pubchem22675
InChI Key RDFJBDMCBVSCFI-UHFFFAOYSA-N
SMILES CN1CCN(CC1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
IUPAC Name 1-methyl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazine
Molecular Weight (g/mol) 302.23
PubChem CID 17998905
Formula Weight 302.22
Percent Purity 97%
Chemical Name or Material 4-(4-Methylpiperazin-1-yl)phenylboronic acid, pinacol ester
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'