missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Methylenebis(N-phenylmaleimide), 95%, Thermo Scientific Chemicals

Product Code. p-7046030
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11313416 5 g 5g
11323416 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11313416 Supplier Thermo Scientific Alfa Aesar Supplier No. L12862.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

4,4'-Methylenebis(N-phenylmaleimide) was used to cure glycidyl ester of eleostearic acid (GEEA).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Methylenebis(N-phenylmaleimide) was used to cure glycidyl ester of eleostearic acid (GEEA).

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13676-54-5
Molecular Formula C21H14N2O4
Molecular Weight (g/mol) 358.353
MDL Number MFCD00005507
InChI Key XQUPVDVFXZDTLT-UHFFFAOYSA-N
Synonym bismaleimide, bismaleimide s, bis 4-maleimidophenyl methane, diphenylmethanebismaleimide, p,p'-dimaleimidodiphenylmethane, 4,4'-methylenebis n-phenylmaleimide, bis p-maleimidophenyl methane, 4,4'-dimaleimidodiphenylmethane, 4,4'-diphenylmethanedimaleimide, 4,4'-biphenylmethanebismaleimide
PubChem CID 83648
IUPAC Name 1-[4-[[4-(2,5-dioxopyrrol-1-yl)phenyl]methyl]phenyl]pyrrole-2,5-dione
SMILES C1=CC(=CC=C1CC2=CC=C(C=C2)N3C(=O)C=CC3=O)N4C(=O)C=CC4=O

Specifications

Melting Point 156°C to 158°C
Quantity 5 g
UN Number UN2811
Beilstein 333986
Formula Weight 358.36
Percent Purity 95%
Chemical Name or Material 4,4'-Methylenebis(N-phenylmaleimide)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.