missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluorophenoxy)benzylamine hydrochloride, 97%, Thermo Scientific™

Product Code. p-7048063
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11359006

Brand: Thermo Scientific Alfa Aesar L20181.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 568565-86-6
Molecular Formula C13H13ClFNO
Molecular Weight (g/mol) 253.701
MDL Number MFCD01862521
InChI Key FSCHAOUFEVMQOV-UHFFFAOYSA-N
Synonym 4-4-fluorophenoxy benzylamine hydrochloride, 4-4-fluorophenoxy phenyl methanamine hydrochloride, 1-4-4-fluorophenoxy phenyl methanamine hydrochloride, acmc-20amw2, 4-4-fluorophenoxy-benzylamine hydrochloride, 4-4-fluorophenoxyl benzylamine hydrochloride, 4-4-fluoro-phenoxy-benzylamine hydrochloride, 1-4-4-fluorophenoxy phenyl methanamine-hydrogen chloride 1/1
PubChem CID 10308088
IUPAC Name [4-(4-fluorophenoxy)phenyl]methanamine;hydrochloride
SMILES C1=CC(=CC=C1CN)OC2=CC=C(C=C2)F.Cl

Specifications

Melting Point 238°C to 240°C
Quantity 1 g
Formula Weight 253.7
Percent Purity 97%
Chemical Name or Material 4-(4-Fluorophenoxy)benzylamine hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.