Learn More
4,4-Difluorocyclohexanone, Maybridge
Product Code. 10402390
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
Product Code. Quantity Packaging unitSize
10402390 1 g Amber Glass Bottle 1g
1 options
Product Code. 10402390
Brand: Thermo Scientific Maybridge MAY00226DA
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
CAS 22515-18-0
Packaging Amber Glass Bottle
Molecular Formula C6H8F2O
Quantity 1 g
Synonym 4,4-difluorocyclohexanone, 4,4-difluoro-cyclohexanone, cyclohexanone, 4,4-difluoro, difluorocyclohexanone, acmc-1cnv9, 4,4-difluoro cyclohexanone, 4,4-difluoro-1-cyclohexanone, 4,4-difluorocycloxanone, 1,1,-difluoro-4-oxocyclohexane
InChI Key NYYSPVRERVXMLJ-UHFFFAOYSA-N
SMILES C1CC(CCC1=O)(F)F
IUPAC Name 4,4-difluorocyclohexan-1-one
Molecular Weight (g/mol) 134.126
PubChem CID 15789092
Formula Weight 134.13
Percent Purity 97%
Chemical Name or Material 4, 4-Difluorocyclohexanone