Learn More
4-(4-Chlorophenyl)-4-oxobutanoic Acid (3-(4-Chlorobenzoyl)propionic Acid), Mikromol™
Product Code. 30050220
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Product Code. 30050220
Brand: Mikromol MM0918.050025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-(4-Chlorophenyl)-4-oxobutanoic Acid (3-(4-Chlorobenzoyl)propionic Acid)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-(4-Chlorophenyl)-4-oxobutanoic Acid (3-(4-Chlorobenzoyl)propionic Acid)
CAS 3984-34-7
Analyte or Component Names 4-(4-Chlorophenyl)-4-oxobutanoic Acid (3-(4-Chlorobenzoyl)propionic Acid)
Shipping Condition Room Temperature
Molecular Formula C10 H9 Cl O3
Certifications/Compliance ISO 17025
Synonym 4-(4-Chlorophenyl)-4-oxobutanoic Acid, 3-(4-Chlorobenzoyl)propionic Acid
InChI Formula InChI=1S/C10H9ClO3/c11-8-3-1-7(2-4-8)9(12)5-6-10(13)14/h1-4H,5-6H2,(H,13,14)
SMILES OC(=O)CCC(=O)c1ccc(Cl)cc1
IUPAC Name 4-(4-chlorophenyl)-4-oxobutanoic acid
Molecular Weight (g/mol) 212.63
Quantity 25 mg
Formula Weight 212.024 g/mol