Learn More
4-(4-Bromophenoxy)benzaldehyde, 97%, Thermo Scientific Chemicals
Catalog Number 15464317
Click to view available options
Quantity:
1 g
5 g
69240-56-8
C13H9BrO2
277.117
MFCD08276811
WMDDJOBNAOVSLP-UHFFFAOYSA-N
4-4-bromophenoxy benzaldehyde, 4-4-bromo-phenoxy benzaldehyde, 4-4-bromo-phenoxy-benzaldehyde, acmc-20amwk
10636240
4-(4-bromophenoxy)benzaldehyde
C1=CC(=CC=C1C=O)OC2=CC=C(C=C2)Br
69240-56-8
C13H9BrO2
277.117
MFCD08276811
WMDDJOBNAOVSLP-UHFFFAOYSA-N
4-4-bromophenoxy benzaldehyde, 4-4-bromo-phenoxy benzaldehyde, 4-4-bromo-phenoxy-benzaldehyde, acmc-20amwk
10636240
4-(4-bromophenoxy)benzaldehyde
C1=CC(=CC=C1C=O)OC2=CC=C(C=C2)Br
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 69°C to 73°C
Quantity 1 g
UN Number UN3077
Sensitivity Air sensitive
Formula Weight 277.12
Percent Purity 97%
Chemical Name or Material 4-(4-Bromophenoxy)benzaldehyde