Learn More
4,4′-Bis(dimethylamino)-benzophenone, Sigma-Aldrich™
Specifications
Specifications
| CAS | 90-94-8 |
| Melting Point | 172°C to 175°C |
| Molecular Formula | [(CH3)2N]C6H4COC6H4-4-[N(CH3)2] |
| MDL Number | MFCD00008312 |
| Quantity | 250 g |
| Synonym | 4,4′-Bis(dimethylamino)-benzophenone, N,N,N,N′-Tetramethyl-4,4′-diaminobenzophenone, Michler ketone, 4,4′-Bis(dimethylamino)-benzophenone |
| Molecular Weight (g/mol) | 268.35 |
| Grade | Research |