Learn More
4,4,5,5-Tetramethyl-2-[3-(phenoxymethyl)phenyl]-1,3,2-dioxaborolane, ≥97%, Thermo Scientific™
Product Code. 10728835
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10728835 250 mg Amber Glass Bottle 250mg
10002214 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10728835 Supplier Thermo Scientific Maybridge Supplier No. CC63639CB
 Restricted Item
Estimated Shipment: 04-12-2026
Chemical Identifiers
CAS 912569-68-7
Molecular Formula C19H23BO3
Molecular Weight (g/mol) 310.20
MDL Number MFCD09065022
InChI Key AFDZGEOQKHIWKF-UHFFFAOYSA-N
Synonym 4,4,5,5-tetramethyl-2-3-phenoxymethyl phenyl-1,3,2-dioxaborolane
PubChem CID 24229702
IUPAC Name 4,4,5,5-tetramethyl-2-[3-(phenoxymethyl)phenyl]-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC(COC2=CC=CC=C2)=CC=C1
Specifications
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97+%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 310.2
Percent Purity 97%
Chemical Name or Material 4,4,5,5-Tetramethyl-2-[3-(phenoxymethyl)phenyl]-1,3,2-dioxaborolane