Learn More
4-{[4-(4-Methyloxy-phenyl)-piperazin-1-yl]-phenyl}-carbamic Acid Phenyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | 4-{[4-(4-Methyloxy-phenyl)-piperazin-1-yl]-phenyl}-carbamic acid phenyl ester |
| CAS | 74853-06-8 |
| Molecular Formula | C24 H25 N3 O3 |
| Quantity | 100 mg |
| Synonym | Itraconazole Methoxy Phenoxy Impurity |
| InChI Formula | InChI=1S/C24H25N3O3/c1-29-22-13-11-21(12-14-22)27-17-15-26(16-18-27)20-9-7-19(8-10-20)25-24(28)30-23-5-3-2-4-6-23/h2-14H,15-18H2,1H3,(H,25,28) |
| SMILES | COc1ccc(cc1)N2CCN(CC2)c3ccc(NC(=O)Oc4ccccc4)cc3 |
| IUPAC Name | phenyl N-[4-[4-(4-methoxyphenyl)piperazin-1-yl]phenyl]carbamate |
| Molecular Weight (g/mol) | 403.47 |
| Formula Weight | 403.1896 |