Learn More
4-(3-Trifluoromethylbenzoyl)piperidine Hydrochloride, TRC
Product Code. 30455370
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30455370 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30455370 Supplier Toronto Research Chemicals Supplier No. TRCT79117050MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-(3-Trifluoromethylbenzoyl)-piperidine','Hydrochloride
CAS 64670-97-9
Molecular Formula C13 H14 F3 N O . Cl H
Quantity 50 mg
InChI Formula InChI=1S/C13H14F3NO.ClH/c14-13(15,16)11-3-1-2-10(8-11)12(18)9-4-6-17-7-5-9;/h1-3,8-9,17H,4-7H2;1H
SMILES Cl.FC(F)(F)c1cccc(c1)C(=O)C2CCNCC2
IUPAC Name piperidin-4-yl-[3-(trifluoromethyl)phenyl]methanone;hydrochloride
Molecular Weight (g/mol) 293.71
Formula Weight 293.0794