missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Thienyl)benzoic acid, 97%, Thermo Scientific™

Product Code. p-2700615
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10252932

Brand: Thermo Scientific Maybridge MO07004CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 29886-64-4
Molecular Formula C11H8O2S
Molecular Weight (g/mol) 204.243
MDL Number MFCD03783559
InChI Key FISAUHGRILVMDP-UHFFFAOYSA-N
Synonym 4-3-thienyl benzoic acid, 4-thiophen-3-yl benzoic acid, 4-thiophen-3-yl-benzoic acid, benzoic acid, 4-3-thienyl, 4-thien-3-ylbenzoic acid, 4-3-thienyl-benzoic acid, maybridge3_004545, acmc-1cisg
PubChem CID 736862
IUPAC Name 4-thiophen-3-ylbenzoic acid
SMILES C1=CC(=CC=C1C2=CSC=C2)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 204.25
Percent Purity 97%
Chemical Name or Material 4-(3-Thienyl)benzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.