Learn More
4-(2′-Pyridylazo)-resorcinol, Sigma-Aldrich™
Description
for synthesis
Specifications
Specifications
| CAS | 1141-59-9 |
| MDL Number | MFCD00006256 |
| Quantity | 5 g |
| Synonym | 4-(2′-Pyridylazo)-resorcinol, 4-(2-Pyridinylazo)-1,3-dihydroxybenzene, 4-(2′-Pyridylazo)-resorcinol |
| Molecular Weight (g/mol) | 215.21 |
| Grade | Research |