Learn More
4-(2-Fluoro-4-nitrophenyl)morpholine, TRC
Product Code. 30405230
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30405230 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30405230 Supplier Toronto Research Chemicals Supplier No. TRCF59486010G
 Restricted Item
Estimated Shipment: 24-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-(2-Fluoro-4-nitrophenyl)morpholine
CAS 2689-39-6
Molecular Formula C10 H11 F N2 O3
Quantity 10 g
Synonym N-(2-Fluoro-4-nitrophenyl)morpholine
InChI Formula InChI=1S/C10H11FN2O3/c11-9-7-8(13(14)15)1-2-10(9)12-3-5-16-6-4-12/h1-2,7H,3-6H2
SMILES [O-][N+](=O)c1ccc(N2CCOCC2)c(F)c1
IUPAC Name 4-(2-fluoro-4-nitrophenyl)morpholine
Molecular Weight (g/mol) 226.2
Formula Weight 226.0754