Learn More
4-[2-(Cyclopropylmethoxy)ethyl]phenol, Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
4-[2-(Cyclopropylmethoxy)ethyl]phenol
Specifications
Specifications
| Chemical Name or Material | 4-[2-(Cyclopropylmethoxy)ethyl]phenol |
| CAS | 63659-16-5 |
| Analyte or Component Names | 4-[2-(Cyclopropylmethoxy)ethyl]phenol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H16 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Betaxolol Hydrochloride Imp. D (EP), Betaxolol Imp. D (EP), 4-[2-(Cyclopropylmethoxy)ethyl]phenol, Betaxolol Hydrochloride Impurity D, Betaxolol Impurity D |
| InChI Formula | InChI=1S/C12H16O2/c13-12-5-3-10(4-6-12)7-8-14-9-11-1-2-11/h3-6,11,13H,1-2,7-9H2 |
| SMILES | Oc1ccc(CCOCC2CC2)cc1 |
| IUPAC Name | 4-[2-(cyclopropylmethoxy)ethyl]phenol |
| Show More |