Learn More
4-(2-Bromophenyl)-2-methyl-1,3-thiazole, 97%, Thermo Scientific™
Product Code. 10336284
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10336284 1 g Amber Glass Bottle 1g
10142984 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10336284 Supplier Thermo Scientific Maybridge Supplier No. CC68410DA
 Restricted Item
Estimated Shipment: 27-11-2026
Chemical Identifiers
CAS 941717-01-7
Molecular Formula C10H8BrNS
Molecular Weight (g/mol) 254.15
MDL Number MFCD09879981
InChI Key SXJQVZDDCODIST-UHFFFAOYSA-N
Synonym 4-2-bromophenyl-2-methyl-1,3-thiazole, 4-2-bromophenyl-2-methylthiazole
PubChem CID 24229764
IUPAC Name 4-(2-bromophenyl)-2-methyl-1,3-thiazole
SMILES CC1=NC(=CS1)C1=CC=CC=C1Br
Specifications
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 254.15
Percent Purity 97%
Chemical Name or Material 4-(2-Bromophenyl)-2-methyl-1, 3-thiazole