missing translation for 'onlineSavingsMsg'
Learn More

4-[2-(3-Pyridyl)-4-thiazolyl]benzeneboronic acid, 97%, Thermo Scientific™

Catalog Number 15473956
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1449132-91-5
C14H11BN2O2S
282.124
MFCD22682932
BRRUQLJIXNIGPZ-UHFFFAOYSA-N
4-2-3-pyridyl-4-thiazolyl benzeneboronic acid, 4-2-pyridin-3-yl-1,3-thiazol-4-yl phenylboronic acid
73996295
[4-(2-pyridin-3-yl-1,3-thiazol-4-yl)phenyl]boronic acid
B(C1=CC=C(C=C1)C2=CSC(=N2)C3=CN=CC=C3)(O)O
This item is not returnable. View return policy
1449132-91-5
C14H11BN2O2S
282.124
MFCD22682932
BRRUQLJIXNIGPZ-UHFFFAOYSA-N
4-2-3-pyridyl-4-thiazolyl benzeneboronic acid, 4-2-pyridin-3-yl-1,3-thiazol-4-yl phenylboronic acid
73996295
[4-(2-pyridin-3-yl-1,3-thiazol-4-yl)phenyl]boronic acid
B(C1=CC=C(C=C1)C2=CSC(=N2)C3=CN=CC=C3)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 282.12
Percent Purity 97%
Chemical Name or Material 4-[2-(3-Pyridyl)-4-thiazolyl]benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'