Learn More
4'-[(1,7'-Dimethyl-2'-propyl[2,5'-bi-1H-benzimidazol]-1'-yl)methyl][1,1'-biphenyl]-2-carboxylic Acid, TRC
Product Code. 18092440
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18092440 10 mg 10mg
18074030 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18092440 Supplier Toronto Research Chemicals Supplier No. TRCD29304510MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1026353-20-7
Molecular Formula C33 H30 N4 O2
Molecular Weight (g/mol) 514.62
Synonym 4'-[(1,7'-Dimethyl-2'-propyl[2,5'-bi-1H-benzimidazol]-1'-yl)methyl][1,1'-biphenyl]-2-carboxylic acid, 4'-[(1,7'-dimethyl-2'-propyl-1H,1'H-2,5'-bibenzo[d]imidazol-1'-yl)methyl]biphenyl-2-carboxylic acid, Telmisartan Imp. B (EP), Telmisartan RC B (USP)
IUPAC Name 2-[4-[[7-methyl-5-(1-methylbenzimidazol-2-yl)-2-propylbenzimidazol-1-yl]methyl]phenyl]benzoic acid
SMILES CCCc1nc2cc(cc(C)c2n1Cc3ccc(cc3)c4ccccc4C(=O)O)c5nc6ccccc6n5C
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C33H30N4O2/c1-4-9-30-34-28-19-24(32-35-27-12-7-8-13-29(27)36(32)3)18-21(2)31(28)37(30)20-22-14-16-23(17-15-22)25-10-5-6-11-26(25)33(38)39/h5-8,10-19H,4,9,20H2,1-3H3,(H,38,39)
Formula Weight 514.2369
Percent Purity >95%
Chemical Name or Material 4'-[(1,7'-dimethyl-2'-propyl-1H,1'H-2,5'-bibenzo[d]imidazol-1'-yl)methyl]biphenyl-2-carboxylic acid
Grade HPLC