Learn More
UVP 3UV™ Lamp
Click to view available options
Wavelength:
365/302/254 nm
Unit Size:
Each
Description
- Three wavelengths, 254/302/365nm UV, in one lamp
- Selection of each wavelength is by an easy turn of the dial located on one end of the housing
- The uniquely designed reflector surfaces mounted behind each tube provides maximum UV for fluorescence applications
- Lightweight and ergonomically designed for hand or stationary use
- Design allows the stationary use by standing the lamp on its end or side
- Available in four, six or eight watts
- Applications:
- Inspection
- Laboratory
- Education
Specifications
Specifications
| Voltage | 230 V |
| Wattage | 8 W |
| Wavelength | 365/302/254 nm |
| Dimensions (L x W x H) | 394 x 76 x 114 mm |
| Electrical Requirements | 8 W, 230 V, 50 Hz |
| Hertz | 50 Hz |
| Model | 3UV-38 |
| Product Type | 3UV Lamp |