Description
- Provides a comfortable respiratory solution for workers in cold and challenging environments.
- Heats compressed air by up to 28°C to enhance user comfort.
- Compatible with a wide range of 3M headtops, including hoods and helmets.
- Allows easy configuration based on environment, contaminants, and personal preferences.
- Features adjustable airflow control to customize heating levels.
- Suitable for use in manufacturing, construction, and other demanding industries.
- Helps improve worker safety and overall comfort during extended use.
Specifications
Specifications
| Color | Black |
| Product Type | Heating Valve Assembly |
| Size | One Size |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?