Learn More
3M™ Versaflo™ M-100 Series PAPR Accessory, Forehead Seal
Product Code. 11727424 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Versaflo M-100 Series PAPR Systems
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11727424 Versaflo M-100 Series PAPR Systems Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11727424 Supplier 3M™ Supplier No. 7100063889
  • ONLINE EXCLUSIVE PRICE
    521.00 NOK
    341.87 NOK / Pack of 1
    Save 179.13 NOK
    34% Off
Estimated Shipment: 13-01-2027
Forehead seal is a replacement part for Versaflo™ M-100 Series faceshields.
  • 3M Catalog# M-154
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Disposable Reusable
Product Type Forehead Seal
For Use With (Application) Cleaning, Electrical, Facility Maintenance, Painting, Sanding, Welding
For Use With (Equipment) Versaflo M-100 Series PAPR Systems
Model M-154