Learn More
3M™ Versaflo™ M-100 Series PAPR Accessory, Airflow Deflector
Product Code. 11797414 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Versaflo M-100 Series PAPR Systems
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11797414 Versaflo M-100 Series PAPR Systems Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11797414 Supplier 3M™ Supplier No. 7100063888
  • ONLINE EXCLUSIVE PRICE
    1220.00 NOK
    800.75 NOK / Pack of 1
    Save 419.25 NOK
    34% Off
 Restricted Item
Estimated Shipment: 01-02-2027
Airflow deflector is a replacement part for Versaflo™ M-100 Series faceshields.
  • 3M Catalog# M-116
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Disposable Reusable
Product Type Airflow Deflector
For Use With (Application) Cleaning, Electrical, Facility Maintenance, Painting, Sanding, Welding
For Use With (Equipment) Versaflo M-100 Series PAPR Systems
Model M-116