Learn More
3M™ Qualitative Fit-Test Kit Accessories
Description
- Test kits and accessories designed to help your organization meet OSHA fit testing requirements for respirators
- The products can be used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate prefilter
- 3M Catalog# 601, FT-13, FT-14, FT-15
Specifications
Specifications
| Product Type | Collar |
| Certifications/Compliance | OSHA |
| For Use With (Application) | Provide secure hood environment |
| For Use With (Equipment) | Qualitative fit test apparatus FT10 and FT30 |
| Model | FT-15 |