Learn More
3M™ TR-300 and TR-600 PAPR Battery Packs
Description
- Rechargeable battery packs
- Lithium Ion
- Long run time for filters and cartridges
Battery packs must be used in accordance with the user instructions, warnings and limitations accompanying each product
Compliance
NIOSH approved.
Specifications
Specifications
| Battery Life | 8 to 18 hr. |
| Battery Type | Lithium-ion |
| Color | Blue, Gray |
| Product Type | Battery Pack |
| For Use With (Application) | Supplying power for PAPR systems |
| For Use With (Equipment) | Versaflo TR-600 PAPR Series |
| Model | TR-632 |
| Quantity | 1/Cs. |