Learn More
3M™ High-Capacity Chemical Sorbents
Click to view available options
Capacity (English):
10.5 gal.
Length (English):
50 ft.
Width (English):
5 in.
Unit Size:
Pack of 3
Description
- Suitable for a wide range of liquids
- Lightweight, easy to deploy and dust free
- Identifiable coloring for easy identification and visibility
- Can be wrung out and reused
- High absorption capacity minimizes amount of waste for disposal
- L x W: 50 ft. x 5 in. (15.2m x 12.7cm)
- Sorbing Capacity: 31.5 gal. (119.2L)
- 3M Catalog# CFL550DD , 3M Catalog# CRL15150 , 3M Catalog# PD914
These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
Specifications
Specifications
| Certifications/Compliance | OSHA 29 CFR 1910.132 |
| Color | Yellow |
| Capacity (English) | 10.5 gal. |
| Capacity (Metric) | 39.74 L |
| Material | Polyester |
| Type | High Capacity Chemical Sorbent |
| For Use With (Application) | For use on most aggressive (hazardous) and non-aggressive liquids |
| Model | CFL550DD |
| Dimensions (L x W) | 15.24 m x 12.7 cm (50 ft. x 5 in.) |
| Length (English) | 50 ft. |
| Show More |