Description
3M™ Cap Mount Hard Hat Headgear H18 provides reliable head and face protection products that meet regulated performance and safety requirements. Headgear systems give users a wide choice of faceshield materials, sizes, shapes, and tints for task specific requirements.
- Versatile headgear assembly can be combined with any 3M faceshield for face protection
- Mounts to 3M H-700 Hard Hats via the accessory slots
- Noise Reduction Rating (NRR)* 28 dB. CSA Class AL
- Requires protective eyewear, sold separately
- High-strength thermoplastic cap-mount
- Provides impact resistance and features a unique system that allows window replacement in seconds
- Meets ANSI Z87.1-2010 requirements
- 3M Catalog# 82502-00000
Specifications
Specifications
| Color | Blue |
| Model | 82502-00000 |
| Product Type | Headgear Combination |
| Size | Standard (6.5 to 8 in.) |
| Ventilated | Not Ventilated |