Learn More
3alpha-Hydroxy Tibolone, TRC
Specifications
Specifications
| Chemical Name or Material | 3alpha-Hydroxy Tibolone |
| CAS | 100239-44-9 |
| Molecular Formula | C21 H30 O2 |
| Quantity | 1 mg |
| Synonym | 19-Norpregn-5(10)-en-20-yne-3,17-diol, 7-methyl-, (3α,7α,17α)-, (3α,7α,17α)-7-Methyl-19-norpregn-5(10)-en-20-yne-3,17-diol, 3α-Hydroxy tibolone, 3α-Hydroxytibolone, Org 4094 |
| InChI Formula | InChI=1S/C21H30O2/c1-4-21(23)10-8-18-19-13(2)11-14-12-15(22)5-6-16(14)17(19)7-9-20(18,21)3/h1,13,15,17-19,22-23H,5-12H2,2-3H3/t13-,15-,17-,18+,19-,20+,21+/m1/s1 |
| SMILES | C[C@@H]1CC2=C(CC[C@@H](O)C2)[C@H]3CC[C@@]4(C)[C@@H](CC[C@@]4(O)C#C)[C@H]13 |
| IUPAC Name | (3R,7R,8R,9S,13S,14S,17R)-17-ethynyl-7,13-dimethyl-2,3,4,6,7,8,9,11,12,14,15,16-dodecahydro-1H-cyclopenta[a]phenanthrene-3,17-diol |
| Molecular Weight (g/mol) | 314.46 |
| Formula Weight | 314.2246 |