Learn More
ThermoSafe 377R/281 Dry Ice Storage Chest Dolly
Product Code. 15654584 Shop All Cole-Parmer Products
Change view
Click to view available options
Holds:
1.6, 2.5 cu. ft. Chests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
15654584 1.6, 2.5 cu. ft. Chests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15654584 Supplier ThermoSafe Supplier No. 0372110
Power Your Renewable Energy Research
Use promo code 28029
View all promotions
Estimated Shipment: 02-12-2026
Move your large-capacity storage chests easily on these ThermoSafe aluminum dollies, even when filled to capacity with dry ice.
  • Large swivel casters roll easily
  • Rim on edge of platform keeps containers on dolly during acceleration or bump stops
  • Grey hammertone finish on heavy-gauge aluminum
  • NSF certified
Specifications
Holds 1.6, 2.5 cu. ft. Chests
Load Bearing Capacity (English) 1.6 and 2.5 cu. ft.
Material Aluminum
Caster Diameter (Metric) 10 cm
Color Gray
For Use With (Equipment) 377R/281 Dry Ice Storage Chest