Learn More
Sampling Systems™ 316 Stainless Steel Micro Spatula
Shop All Sampling Systems Products
Click to view available options
Length (Metric):
175 mm
Unit Size:
Each
Description
- Stainless steel
- For powder sampling
- Crevice free
- Bright polish finish
Specifications
Specifications
| Product Type | Micro Spatula |
| Length (Metric) Blade | 40 mm |
| Width (Metric) Blade | 12 mm |
| Material | Stainless Steel |
| Length (Metric) | 175 mm |