Learn More
Thermo Scientific™ 316 Stainless Steel Capillary Tubing for HPLC
Product Code. 10762517 Shop All Thermo Scientific Products
Click to view available options
Overall Length (Metric):
5 cm
10 cm
10.5 cm
15 cm
20 cm
28 cm
30 cm
40 cm
50 cm
100 cm
Diameter (Metric) Inner:
0.127 mm
0.18 mm
0.25 mm
Color:
Black
Blue
Red
Yellow
Unit Size:
Each
Product Code. 10762517
missing translation for 'mfr': Thermo Scientific™ 39011FLEX
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use cleaned, polished, passivated and ready-to-use Thermo Scientific™ 316 Stainless Steel Capillary Tubing for HPLC.
Use Thermo Scientific™ 316 Stainless Steel Capillary Tubing for ultra-high pressure applications. Prefinished, square, burr-free ends and interiors ensure minimum dead volume connections. Cleaned, polished and passivated, it is ready for immediate use. This tubing is not recommended for biological samples.
  • Suitable for ultra high pressure applications
  • Wide chemical compatibilty
  • Prefinished, square, burr-free ends and interiors to minimize dead volume connections
TRUSTED_SUSTAINABILITY
Specifications
For Use With HPLC
Type Capillary Tubing
Color Yellow
For Use With (Application) HPLC
Diameter (English) Inner 0.007 in.
Diameter (Metric) Inner 0.18 mm
Overall Length (English) 4.134 in.
Overall Length (Metric) 10.5 cm
Material Stainless Steel