Learn More
Thermo Scientific™ 316 Stainless Steel Capillary Tubing for HPLC
Product Code. 10531885 Shop All Thermo Scientific Products
Change view
Click to view available options
Overall Length (Metric):
5 cm
10 cm
10.5 cm
15 cm
20 cm
28 cm
30 cm
40 cm
50 cm
100 cm
Diameter (Metric) Inner:
0.127 mm
0.18 mm
0.25 mm
Color:
Black
Blue
Red
Yellow
Unit Size:
Each
Product Code. Overall Length (Metric) Diameter (Metric) Inner Color unitSize
10531885 5 cm 0.127 mm Red Each
10762517 10.5 cm 0.18 mm Yellow Each
10679665 15 cm 0.18 mm Yellow Each
10505425 28 cm 0.18 mm Yellow Each
10641396 40 cm 0.18 mm Yellow Each
10764994 5 cm 0.25 mm Blue Each
10582265 10 cm 0.25 mm Blue Each
10160603 20 cm 0.25 mm Blue Each
10592265 30 cm 0.25 mm Blue Each
10752337 5 cm 0.18 mm Black Each
10370965 10 cm 0.18 mm Black Each
10357214 30 cm 0.18 mm Black Each
10289464 50 cm 0.18 mm Black Each
10025734 100 cm 0.18 mm Black Each
10521885 50 cm 0.25 mm Blue Each
10735374 100 cm 0.25 mm Blue Each
10253703 10 cm 0.127 mm Red Each
10756524 20 cm 0.127 mm Red Each
10150553 30 cm 0.127 mm Red Each
10446113 50 cm 0.127 mm Red Each
10784604 100 cm 0.127 mm Red Each
21 options
Product Code. 10531885
Brand: Thermo Scientific™ U152
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use cleaned, polished, passivated and ready-to-use Thermo Scientific™ 316 Stainless Steel Capillary Tubing for HPLC.
Use Thermo Scientific™ 316 Stainless Steel Capillary Tubing for ultra-high pressure applications. Prefinished, square, burr-free ends and interiors ensure minimum dead volume connections. Cleaned, polished and passivated, it is ready for immediate use. This tubing is not recommended for biological samples.
  • Suitable for ultra high pressure applications
  • Wide chemical compatibilty
  • Prefinished, square, burr-free ends and interiors to minimize dead volume connections
TRUSTED_SUSTAINABILITY
Specifications
For Use With HPLC
Type Capillary Tubing
Color Red
For Use With (Application) HPLC
Diameter (English) Inner 0.005 in.
Diameter (Metric) Inner 0.127 mm
Overall Length (English) 1.969 in.
Overall Length (Metric) 5 cm
Material Stainless Steel