Learn More
Thermo Scientific™ 316 Stainless Steel Capillary Tubing for HPLC in 5-Foot Coils
Shop All Thermo Scientific ProductsDescription
Use Thermo Scientific™ 316 Stainless Steel Capillary Tubing for ultrahigh-pressure applications. Prefinished, square, burr-free ends and interiors ensure minimum dead volume connections. Cleaned, polished and passivated, it is ready for immediate use. This tubing is not recommended for biological samples.
- Suitable for ultra high pressure applications
- Wide chemical compatibilty
Specifications
Specifications
| For Use With | HPLC |
| Type | Capillary Tubing |
| Description | 0.040 in. ID; 5 ft. length |
| For Use With (Application) | HPLC |
| Diameter (English) Inner | 0.040 in. |
| Diameter (Metric) Inner | 1.02 mm |
| Overall Length (English) | 5 ft. |
| Overall Length (Metric) | 1.524 m |
| Material | Stainless Steel |