missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC7 Antibody [FITC], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38013F
This item is not returnable.
View return policy
Description
DNAJC7 Polyclonal antibody specifically detects DNAJC7 in Human samples. It is validated for ELISA,Western Blot
Specifications
| DNAJC7 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 7266 | |
| Store at 4°C in the dark. | |
| Purified |
| ELISA, Western Blot | |
| FITC | |
| DANJC7, DJ11, DnaJ (Hsp40) homolog, subfamily C, member 7, tetratricopeptide repeat domain 2, Tetratricopeptide repeat protein 2, TPR repeat protein 2, TPR2DJC7, TTC2dnaJ homolog subfamily C member 7 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 230-380 of human DNAJC7 (NP_003306.3).,, Sequence:, VQFFVQALRMAPDHEKACIACRNAKALKAKKEDGNKAFKEGNYKLAYELYTEALGIDPNNIKTNAKLYCNRGTVNSKLRKLDDAIEDCTNAVKLDDTYIKAYLRRAQCYMDTEQYEEAVRDYEKVYQTEKTKEHKQLLKNAQLELKKSKRK | |
| 0.1 mL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction