missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tropomyosin-3 Antibody [Janelia Fluor« 646], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35209JF646
This item is not returnable.
View return policy
Description
tropomyosin-3 Polyclonal antibody specifically detects tropomyosin-3 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| tropomyosin-3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 7170 | |
| Store at 4°C in the dark. | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Janelia Fluor 646 | |
| cytoskeletal tropomyosin TM30, FLJ41118, gamma-tropomyosin, heat-stable cytoskeletal protein 30 kDa, hscp30, hTM5, MGC14582, MGC3261, MGC72094, NEM1, OK/SW-cl.5, TM3, TM30, TM30nm, TM-5, TPMsk3, TRK, tropomyosin 3, tropomyosin alpha-3 chain, tropomyosin gamma, tropomyosin-3, Tropomyosin-5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-285 of human tropomyosin-3 (NP_689476.2).,, Sequence:, MMEAIKKKMQMLKLDKENALDRAEQAEAEQKQAEERSKQLEDELAAMQKKLKGTEDELDKYSEALKDAQEKLELAEKKAADAEAEVASLNRRIQLVEEELDRAQERLATALQKLEEAEKAADESERGMKVIENRALKDEEKMELQEIQLKEAKHIAEEADRKYEEVARKLVIIEGDLERTEERAELAESKCSELEEELKNVTNNLKSLEAQAEKYSQKEDKYEEEIKILTDKLKEAETRAEFAERSVAKLEKTIDDLEDELYAQKLKYKAISEELDHALNDMTSI | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction