Learn More
DHX36 Antibody [PerCP], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-27883PCP
Product Code. 30499953
  • 5075.00 NOK / 0.10mL
Description
DHX36 Polyclonal antibody specifically detects DHX36 in Human,Mouse samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DHX36
Polyclonal
PBS
Rabbit
Peptide affinity purified
RUO
170506
Store at 4°C in the dark.
Purified
ELISA, Western Blot, Immunohistochemistry (Paraffin)
PerCP
DDX36, DEAH (Asp-Glu-Ala-His) box polypeptide 36, DEAH box protein 36, EC 3.6.1, EC 3.6.4.13, G4 resolvase-1, KIAA1488G4R1, MLEL1RHAUDEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 36, MLE-like protein 1, probable ATP-dependent RNA helicase DHX36, RNA helicase associated with AU-rich element ARE
DHX36 antibody was raised against a 19 amino acid peptide near the carboxy terminus of human DHX36. The immunogen is located within the last 50 amino acids of DHX36.
0.1 mL
Primary
Human, Mouse