missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ADRM1 Polyclonal antibody specifically detects ADRM1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | ADRM1 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 646 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | 000 surface antigen, 110 kDa cell membrane glycoprotein, adhesion regulating molecule 1, Adhesion-regulating molecule 1, ARM1, ARM-1, Gp110, GP110hRpn13, M(r) 110, MGC29536, proteasomal ubiquitin receptor ADRM1, Proteasome regulatory particle non-ATPase 13, regulatory particle non-ATPase 13, Rpn13, Rpn13 homolog |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 108-407 of human ADRM1 (NP_008933.2).,, Sequence:, WMQEPKTDQDEEHCRKVNEYLNNPPMPGALGASGSSGHELSALGGEGGLQSLLGNMSHSQLMQLIGPAGLGGLGGLGALTGPGLASLLGSSGPPGSSSSSSSRSQSAAVTPSSTTSSTRATPAPSAPAAASATSPSPAPSSGNGASTAASPTQPIQLSDLQSILATMNVPAGPAGGQQVDLASVLTPEIMAPILANADVQERLLPYLPSGESLPQTADEIQNTLTSPQFQQALGMFSAALASGQLGPLMCQFGLPAEAVEAANKGDVEAFAKAMQNNAKPEQKEGDTKDKKDEEEDMSLD |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?