missing translation for 'onlineSavingsMsg'
Learn More

Ext2 Antibody [Janelia Fluor« 549], Novus Biologicals Biologicals™

Product Code. 30498727 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30498727 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30498727 Supplier Novus Biologicals Supplier No. NBP335904JF549
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Ext2 Polyclonal antibody specifically detects Ext2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ext2
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias EC 2.4.1.224, EC 2.4.1.225, exostoses (multiple) 2, exostosin 2, exostosin-2, Glucuronosyl-N-acetylglucosaminyl-proteoglycan/N-acetylglucosaminyl-proteoglycan4-alpha-N-acetylglucosaminyltransferase, Multiple exostoses protein 2, N-acetylglucosaminyl-proteoglycan 4-beta-glucuronosyltransferase, Putative tumor suppressor protein EXT2, SOTV
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 80-340 of human Ext2 (NP_000392.3).,, Sequence:, PHSIESSNDWNVEKRSIRDVPVVRLPADSPIPERGDLSCRMHTCFDVYRCGFNPKNKIKVYIYALKKYVDDFGVSVSNTISREYNELLMAISDSDYYTDDINRACLFVPSIDVLNQNTLRIKETAQAMAQLSRWDRGTNHLLFNMLPGGPPDYNTALDVPRDRALLAGGGFSTWTYRQGYDVSIPVYSPLSAEVDLPEKGPGPRQYFLLSSQVGLHPEYREDLEALQVKHGESVLVLDKCTNLSEGVLSVRKRCHKHQVFD
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 2132
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.